Learn More
BUCHI Condensers for BUCHI Rotavapors
Product Code. 15600655 Shop All Buchi Products
Click to view available options
Type:
Condenser S
Vertical Condenser V
Material (Coating):
P+G
Uncoated
Unit Size:
Each
Product Code. 15600655
Brand: BUCHI 11059368
Only null left
Add to basket
Add to basket
Original BUCHI glassware guarantees the highest levels of efficiency and safety thanks to high precision in manufacturing. For more than 80 years, BUCHI has been manufacturing innovative and durable glass parts to ease daily laboratory work.
  • High throughput due to advanced design of condensers
  • Highly resistant glass provides the highest levels of safety (DURAN™ borosilicate glass 3.3)
  • Guaranteed leak-tightness and protection against hazardous fumes thanks to high precision joints
  • Maximum stability and safety with plastic and glass coating
Specifications
Material Borosilicate Glass (3.3)
Type Vertical Condenser V
For Use With (Equipment) Rotavapor R-210/R-215
Material (Coating) Uncoated