Learn More
Cytiva Conductivity Flow Cell
Product Code. p-8001012 Shop All Cytiva Products
Click to view available options
Material:
Titanium and Chlorotrifluoroethylene
Titanium, Ethylene propylene diene and Fluoro plastics
Unit Size:
Each
Product Code. 11330232
Brand: Cytiva 18111105
Only null left
Specifications
Connection Type UNF 10-32 2 B finger tights
For Use With (Equipment) Monitor pH/C-900
Material Titanium and Chlorotrifluoroethylene
Max. Pressure 50 bar
Type Conductivity Flow Cell
Volume (Metric) 0.041 mL
pH 1 to 14
Quantity 1 Pack
Diameter (Metric) Outer 4.23 mm