missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard 84μS/cm, Hanna Instruments™
Click to view available options
Quantity:
500 mL
Packaging:
FDA Bottle
Light Block Bottles
Specifications
Packaging | FDA Bottle |
Quantity | 500 mL |
Color | Colorless |
Odor | Odorless |
pH | 7 |
Solubility | Soluble |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'