missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 111800μS/cm, NIST Certified, Chem-Lab
Click to view available options
Quantity:
1000 mL
Packaging:
Polyethylene
Description
5°C—74.1mS/cm
10°C—83.2mS/cm
15°C—92.5mS/cm
20°C—102.1mS/cm
25°C—111.8mS/cm
30°C—121.7mS/cm
35°C—131.7mS/cm
40°C—142.1mS/cm
Specifications
Concentration | 1.0M |
CAS | 7447-40-7 |
Packaging | Polyethylene |
Quantity | 1000 mL |
Molecular Formula | ClK |
MDL Number | MFCD00011360 |
InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
SMILES | [Cl-].[K+] |
IUPAC Name | potassium chloride |
Molecular Weight (g/mol) | 74.55 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'