missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 12880μS/cm, NIST Certified, Chem-Lab
Click to view available options
Quantity:
1000 mL
Unit Size:
Each
Description
5°C—8.22mS/cm
10°C—9.33mS/cm
15°C—10.48mS/cm
20°C—11.67mS/cm
25°C—12.88mS/cm
30°C—14.12mS/cm
35°C—15.38mS/cm
40°C—16.67mS/cm
Specifications
Specifications
| Packaging | Polyethylene |
| CAS | 7447-40-7 |
| Density | 1.01g/mL |
| Conductivity | 12880μS/cm |
| Molecular Formula | ClK |
| Concentration | 0.1 M |
| MDL Number | MFCD00011360 |
| InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
| SMILES | [Cl-].[K+] |
| IUPAC Name | potassium;chloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction