Learn More
Lenz Laborglasintrumente Cones
Click to view available options
Joint Size:
NS 40/38
NS 5/13
NS 50/42
NS 55/44
NS 60/46
NS 7/16
NS 71/51
NS 85/55
Unit Size:
Each
Pack of 10
Description
- Acc. to DIN 12249 form B and in addition to standard
- Conical ground cones
- Made of DURAN™ tubing
Specifications
Specifications
| Quantity | 1 Ea. |
| Joint Size | NS 55/44 |
| Thickness (Metric) Wall | 2.5 mm |
| Diameter (Metric) Outer Tube | 50 mm |
| Material | Borosilicate Glass (3.3) |