Learn More
Lenz Laborglasintrumente Cones with Drip Tip
Click to view available options
Joint Size:
NS 14/23
NS 19/26
NS 24/29
NS 29/32
Unit Size:
Pack of 10
Description
- Acc. to DIN 12249 form F
- Conical ground cones
- Made of DURAN™ tubing
Specifications
Specifications
| Joint Size | NS 19/26 |
| Thickness (Metric) Wall | 1.8 mm |
| Diameter (Metric) Outer Tube | 17 mm |
| Material | Borosilicate Glass (3.3) |