Learn More
Lenz Laborglasintrumente Conical Joint Clips Type Safety Clip
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01103819
Additional Details : Weight : 0.50000kg
Product Code. 10550845
  • 34.72€ / Pack of 10
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
Description
Description
  • For conical ground joints
  • Material: POM
  • Height adjustable by variable gear rods
  • For use with sleeves with grip collar
  • Outstanding chemical resistance
  • Operating temperature up to approx. 150 °C
Specifications
Specifications
10/Pk.
Gorund Joint NS 19/26
Blue