Learn More
Applied Biosystems™ Connector Cables for Automated Thermal Cycler (ATC)
Shop All Applied Biosystems ProductsDescription
Includes
1 meter connection cable for Automated Thermal Cycler (ATC)
Used to connect the Automated Thermal Cycler (ATC) block to the power box, this connector is available in a rigid 10-cm, flexible 1-m, or flexible 3-m cable format.
For customers seeking to automate their PCR workflow, the Applied Biosystems Automated Thermal Cycler (ATC) provides the reliability and performance you've come to expect from Applied Biosystems instruments. Small, easy-to-integrate, with a compatible format, our Automated Thermal Cycler enables hands-free PCR results.
In order to better align with lab design and demands, a longer or shorter cable may be utilized to optimize workflow.
Warranty
27 month warranty
Specifications
Specifications
| Format | 1-m connector cable (flexible) |
| For Use With (Equipment) | Automated Thermal Cycler (ATC) |
| Product Type | Connector Cable |
| For Use With (Application) | PCR |
| Max. Heating Rate (Thermal Block) | 2.8°C/sec. (Block), 1.6°C/sec. (Sample) |
| Quantity | 1 cable |