missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liofilchem™ Contact Slide 3
PCA and TTC and Neutralizing / Vogel Johnson Agar, Flex Dip-slide with a selective medium for detection of pathogenic staphylococci and a non selective medium for total bacterial count.
47.81€ - 258.65€
Specifications
Species | Staphylococcus species |
---|---|
Certifications/Compliance | ISO 18593:2004 |
Content And Storage | 10°C to 25°C |
Detects | Bacterial Count, Staphylococci |
Format | Slide |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11974369
|
Liofilchem™
525302 |
20 Slides |
47.81€
Pack of 20 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
11964339
|
Liofilchem™
53530 |
120 Slides |
258.65€
Pack of 120 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
Description
- Contact Slide 3 is a ready-to-use device with two different media coated onto a plastic support used for the microbial monitoring of surfaces and liquids even in the presence of residues of disinfectants. The selective medium allows the isolation and enumeration of coagulase-positive, mannitol-utilizing staphylococci. The other medium is used for enumeration of bacteria.
Specifications
Staphylococcus species | |
10°C to 25°C | |
Slide | |
Contact Slide | |
Gram-positive Organisms, Gram-positive Cocci, Gram-positive Cocci | |
For Bacterial Count and Staphylococci Detection | |
24 to 48hr. | |
Flex |
ISO 18593:2004 | |
Bacterial Count, Staphylococci | |
Italy | |
Non-sterile | |
PCA, TTC, Neutralizing/Vogel Johnson Agar | |
At 36 ± 1°C for 24 to 48 hr. | |
ISO 18593:2004 |