missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liofilchem™ Contact Slide 8
Click to view available options
Quantity:
20 Slides
Unit Size:
Pack of 20
Description
- Contact Slide 8 is a ready-to-use device with two different media coated onto a plastic support used for the microbial monitoring of surfaces and liquids even in the presence of residues of disinfectants. The selective medium allows the isolation and enumeration of coliform bacteria. The other medium is used to obtain the total bacterial count.
Specifications
Specifications
| Certifications/Compliance | ISO 18593:2004 |
| Content And Storage | 10°C to 25°C |
| Detects | Bacterial Count, Coliform |
| Format | Slide |
| Manufacturing Origin | Italy |
| Product Type | Contact Slide |
| Sterility | Non-sterile |
| Target Organism Class | Coliforms, Gram-positive Organisms, Coliforms |
| Formulation | PCA + TTC + Neutralizing/VRBL Agar + Neutralizing |
| For Use With (Application) | For Bacterial Count and Coliform Detection |
| Show More |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction