missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liofilchem™ Contact Slide 9
Click to view available options
Quantity:
120 Slides
20 Slides
Unit Size:
Pack of 120
Pack of 20
Description
- Contact Slide 9 is a ready-to-use device with a medium coated onto a plastic support used for the microbial monitoring of surfaces and liquids. The slide is covered on both sides with PCA and TTC and Neutralizing which allows bacteria to grow as red colonies, even in the presence of residues of disinfectants.
Specifications
Specifications
| Certifications/Compliance | ISO 18593:2004 |
| Content And Storage | 10°C to 25°C |
| Detects | Total Bacterial Count |
| Format | Slide |
| Manufacturing Origin | Italy |
| Product Type | Contact Slide |
| Sterility | Non-sterile |
| Target Organism Class | Gram-positive Organisms |
| Formulation | PCA + TTC + Neutralizing/PCA + TTC + Neutralizing |
| For Use With (Application) | For Total Bacterial Count Detection |
| Show More |
Safety and Handling
Product Identifier
- relyon virkon tablets
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- EUH208-Contains . May produce an allergic reaction.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : pentapotassium bis(peroxymonosulphate) bis Sulphate, malic acid, sulphamidic acid, benzenesulphonic
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction