Learn More
Gilac™ Container with Integrated Lid
Click to view available options
Dimensions (L x W x H):
400 x 300 x 310 mm
600 x 400 x 315 mm
Unit Size:
Each
Description
- Container with integrated ''crocodile'' lid.
- Ideal for catering.
- Stackable when closed and nesting when the lid is open.
Specifications
Specifications
| Type | Storage Tote |
| Material | HDPE |
| Color | Blue |
| Shape | Rectangular |
| Capacity (English) | 6.6 gal. |
| Capacity (Metric) | 25 L |
| Dimensions (L x W x H) | 400 x 300 x 310 mm |