Learn More
Contec™ Amplitude™ Lambda Wipes
BIOPHARMAGREENER_CHOICE
Product Code. p-4578311 Shop All Contec Products
Click to view available options
Dimensions (L x W):
9 x 9 in. (22.86 x 22.86 cm)
Unit Size:
Pack of 3600
Product Code. 15616986
Brand: Contec™ AMLA0001
Only null left
Add to basket
Add to basket
Composed of hydraulically entangled blend of 58% cellulose and 42% polyester wipe. Contec™ Amplitude™ Lambda Wipes are strong and durable even when wet.
  • Excellent all-purpose wipes
  • Nonwoven strong and durable hydroentangled fabric
  • Low levels of particles and residues
  • Outstanding sorbency
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Wipe
Length (English) 9 in.
Length (Metric) 23 cm
Width (English) 9 in.
Width (Metric) 23 cm
Material Polyester Cellulose
Dimensions (L x W) 9 x 9 in. (22.86 x 22.86 cm)
For Use With (Application) General purpose cleaning in biotech, pharmaceutical, semiconductor, electronics, medical devices, and printed circuit board industries