Learn More
Contec™ ProZorb™ Nonwoven Cleanroom Wipes
BIOPHARMAGREENER_CHOICE
Product Code. p-210598 Shop All Contec Products
Click to view available options
Dimensions (L x W):
12 x 12 in.
Unit Size:
Pack of 1800
Product Code. 15691393
Brand: Contec™ PZ1212
Only null left
Add to basket
Add to basket
Manufactured with cellulose and polyester through a special process that creates apertures in the fabric while enhancing the thickness. Contec™ ProZorb™ Nonwoven Cleanroom Wipes provide a textured surface with excellent sorptive capacity and particle removal capability.
  • Textured polyester/cellulose nonwoven wipes promotes effective cleaning
  • Low levels of particles and extractables
  • Strong and durable
  • Designed for nonabrasive applications in the biotech, pharmaceutical, electronics, semiconductor industries and more
  • Recommended for use in ISO Class 5 cleanroom applications; Use for product wipe down prior to cleanroom entry 
  • Great for spill clean up in critical environments
  • No chemical binders or surfactants
Order Info
Basis weight: 55g/m2.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Wiper
Length (English) 12 in.
Length (Metric) 30.5 cm
Width (English) 12 in.
Width (Metric) 30.48 cm
Material Polyester Cellulose
Basis Weight 55
Certifications/Compliance ISO Class 5
Dimensions (L x W) 12 x 12 in.
For Use With (Application) Cleanroom