missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Continuous Colored Labels

Continuous Colored Labels, a colored, printable vinyl film for Thermal Transfer printers.

Brand:  Brady™ 217529

Additional Details : Weight : 0.61300kg

Product Code. 11588694

  • 247.00€ / 1 roll
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Colored, printable vinyl film for Thermal Transfer printers.
Specifications

Specifications

Thermal Transfer Printable Labels
White
Vinyl
Labels
BBP11, BBP12, PR Plus, THT 76 mm Core Printers
-70°C
1 Ea.
-70°C to +80°C
0.107 mm
131 ft.
2.48 in.
25.4 mm I.D.
Permanent Acrylic, Permanent
Satin
Thermal Transfer
Barcode labelling, Component and equipment labelling, Data and telecommunications labelling, Electrical labelling, General Identification, Inventory and inspection labelling, Laboratory labelling
1
1 Roll
-94°F to +176°F
0.0042 in.
40 m x 63 mm
40 m
63 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions