Learn More
Scientific Industries Control Board
Product Code. 15229858
Change view
Click to view available options
For Use With (Equipment):
Vortex-Genie 2T
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15229858 Vortex-Genie 2T Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15229858 Supplier Scientific Industries Supplier No. EBP0001
Only null left
Add to basket
Add to basket
Replacement part for the Vortex-Genie 2T Timer Genie.
Replacement part for the Vortex-Genie 2T Timer Genie
Specifications
Product Type Control Board
Color Green
Description Control Board Replacement part
For Use With (Equipment) Vortex-Genie 2T
Includes Replacement board
Packaging Quantity 1 Ea.