Learn More
Linker Industrietechnik™ Control Screen Certificates
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
Sieves (100 mm dia.)
Sieves (200 mm dia.)
Sieves (400 mm dia.)
missing translation for 'unitSize'
Each
Description
- Guarantee of reliability
- Numbered control certificate
- Video measurements carried out on the screen
Specifications
Specifications
| For Use With (Equipment) | Sieves (400 mm dia.) |
| Product Type | Control Screen Certificate |
| Includes | Serial number, manufacturing standard, mesh size and its parameters, the wire diameter . Provide a certificate by Stainless Steel sieve |