Learn More
Plas Labs™ Controlled Purge System
Product Code. 11847142
Click to view available options
Height (Metric):
179 mm
Unit Size:
Each
Product Code. 11847142
Brand: Plas Labs™ 800CPS/EXP
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Product Type Controlled Purge System
Height (Metric) 179 mm
Includes Programmable logic controller, pressure sensor, black Noryl™ housing, flow meter, solenoid valve, and push-to-seal fittings
Quantity 1 Ea.