Learn More
Miele Professional™ Conversion kit
Shop All Miele Products
Click to view available options
:
conversion kit; upper baskets; model G 78 to PG 85; welded water supply
Description
- For older load carriers without dryer connection
- For older load carriers with welded water supply
Specifications
Specifications
| Color | Black |
| Holds | Plastic |