Learn More
3M™ Protective Coverall 4532+, Cool White
Product Code. 15760048
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Unit Size:
Pack of 20
Product Code. 15760048
Brand: 3M™ 7100016012
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Coverall has a unique specialist material treatment which offers enhanced liquid hold out with added oil and alcohol repellency. Certified to Type 5 for hazardous particles and Type 6 for limited chemical splash, 4532+ coverall provides soft breathable comfort of an SSMMS with the improved liquid performance normally associated with a laminate.
  • Unique liquid repellency with oil and alcohol resistance material treatment
  • Soft and strong multi-layered 52gsm SSMMS for strong performance
  • Made from super breathable SSMMS non-woven for effective release of body heat and moisture and reduced heat stress
  • 3 panel hood, reinforced triangular gusset panel, 2-way zip and storm flap with sealable tape, elasticated back and ankles to better freedom of movement
  • Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
  • EN 1073-2 nuclear particulate hold out
  • Knitted cuffs and elastic waist and ankles for a better fit and freedom of movement
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protective Coveralls
Size Large
Gender Unisex
Material SSMMS
Color White
Disposable Single-use
Waist Style Elastic
Cuff Style Knit
Closure Type Front Zipper
Sterility Non-sterile
Certifications/Compliance CE Category III , EN 1073-2 radioactive particles , EN 1149-5 - electrostatic properties , EN 13034: 2005 + A1: 2009 Type 6 liquid splash , EN ISO 13982-1: 2004 + A1: 2010 Type 5 harmful particles
Certifications/Compliance (Europe) CE Category III, EN 1073-2, EN 1149-5, EN 13034:2005+A1:2009 Type 6, EN ISO 13982-1:2004 Type 5