Learn More
VACUUBRAND™ Cooling Water Valve
Click to view available options
Voltage:
24 V
230 V
Plug Type:
Diode Plug
IEC Plug
Unit Size:
Each
Specifications
Specifications
| For Use With | For CVC 2 and VARIO Pumps |
| Plug Type | Diode Plug |
| Quantity | 1 Each |
| Type | Cooling Water Valve |
| Voltage | 24 V |