Learn More
Novus Biologicals™ COOMASSIEnano Protein Staining Solution
Description
COOMASSIE nano, enhanced by nano-technology, is a ready-to-use protein staining solution for SDS-PAGE gels. Its next generation formula offers a faster protein detection, higher sensitivity and there is no need for destaining. Also, the washing step can be omitted. In the absence of hazardous substances such as methanol and acetic acid, COOMASSIE nano is considered to be safe and environmentally friendly. COOMASSIE nano is also compatible with mass spectrophotometry.
Specifications
Specifications
| For Use With (Application) | There is no risk of excessive staining. Instead, superior visibility for each band is achieved when the gel is submerged overnight. Carefully mix COOMASSIE nano up and down few times before use. If the gel is thicker than mini-gel (1mm), it will take longer to incubate. The staining solution may contain blue clumps. This is normal and easily dissolved in distilled water |
| Quantity | 500 mL |