missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Aluminum Bronze, CDA 623, C62300, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Copper, Aluminum Bronze, CDA 623, C62300, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Cobalt/Magnesium/Silver/Carbon/Chromium/Zinc/Silicon/Tin/Manganese/Nickel/Iron/Aluminium/Copper/Lead |
| CAS | 7440-50-8 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Cobalt 0.0003 %/Magnesium 0.0004 %/Silver 0.0009 %/Carbon 0.0028 %/Chromium 0.0028 %/Zinc 0.006 %/Silicon 0.017 %/Tin 0.0172 %/Manganese 0.151 %/Nickel 0.234 %/Iron 2.09 %/Aluminium 9.37 %/Copper 88.06 %/Lead (0.004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction