missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Fluka™ Copper(I) Chloride, ≥97.0% (RT), Honeywell™

Puriss. p.a., ACS Reagent,≥97.0% (RT)

Brand:  Honeywell Fluka™ 61168-250G

Additional Details : CAS : 7758-89-6 Poids : 0.25000kg Transport : UN number : 2802 Chem class : 8 Pack group : III

Packaging Poly bottle
Quantity 250g

 View more versions of this product

Product Code. 15646020

  • 97.33€ / 250g


Expédition estimée
12-03-2021
Add to basket

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7758-89-6
99.00
OXBLHERUFWYNTN-UHFFFAOYSA-M
24593
[Cl-].[Cu+]
ClCu
MFCD00010969
copper i bromide, copper bromide cubr, copper 1+ bromide, copper monobromide, cuprousbromide, hsdb 270, cubr, copper 1 bromide
λ¹-copper(1+) chloride

Specifications

Copper(I) bromide
ClCu
MFCD00010969
copper i bromide, copper bromide cubr, copper 1+ bromide, copper monobromide, cuprousbromide, hsdb 270, cubr, copper 1 bromide
[Cl-].[Cu+]
99.00
143.45g/mol
4.98g/cm3
Poly bottle
1345°C (1013hPa)
492°C
7758-89-6
CuBr
NONH for all modes of transport
OXBLHERUFWYNTN-UHFFFAOYSA-M
λ¹-copper(1+) chloride
24593
≥98.0% (RT)
Odorless
Powder
Colorless
250g
Videos
SDS Documents