missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Oxide, 97%, Honeywell Fluka™

Catalog Number p-7120344
Click to view available options
Quantity:
25 g
500 g
2 kg
12 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
1317-39-1
MFCD00010974
This item is not returnable. View return policy
1317-39-1
MFCD00010974

Powder, <5 μm, 97%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 1230°C
Color Brown or Red
pH 8.0 (at 100g/L 20°C)
Physical Form Powder
Quantity 25 g
Linear Formula Cu2O
UN Number UN3077
Formula Weight 143.09g/mol
Percent Purity 97%
Packaging Poly Bottle
Density 6g/mL at 25°C
Chemical Name or Material Copper(I) Oxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'