missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) chloride hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048500
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
10125-13-0
Cl2CuH4O2
170.48
MFCD00149674
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
61482
CHEBI:86318
dichlorocopper;dihydrate
O.O.[Cl-].[Cl-].[Cu++]
This item is not returnable. View return policy
10125-13-0
Cl2CuH4O2
170.48
MFCD00149674
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
61482
CHEBI:86318
dichlorocopper;dihydrate
O.O.[Cl-].[Cl-].[Cu++]

Copper(II) chloride hydrate promotes the hydrolysis of acetonides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 100°C (decomposition)
Color Blue-Green
pH 3.0 to 3.8
Physical Form Crystalline Aggregates
Quantity 5 g
UN Number UN2802
Merck Index 14,2633
Solubility Information Soluble in water.
Formula Weight 134.45 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Density 2.510 g/mL
Assay (metals basis)
Chemical Name or Material Copper(II) chloride hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'