missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) fluoride hydrate, Puratronic™, 99.99% (metals basis)

Product Code. 11372098
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11372098

Brand: Thermo Scientific Alfa Aesar 013001.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 13454-88-1 | CuF2H4O2 | 137.57 g/mol

Copper(II) fluoride hydrate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-88-1
Molecular Formula CuF2H4O2
Molecular Weight (g/mol) 137.57
MDL Number MFCD00010977
InChI Key CTRICEWDSISGPV-UHFFFAOYSA-L
Synonym cupric fluoride dihydrate, unii-3a38pc42e9, copper ii fluoride dihydrate, copper difluoride, dihydrate, cupric fluoride dihydrate mi, copper fluoride cuf2 , dihydrate, acmc-1bwnw, copper 2+ dihydrate difluoride, copper 2+ ion dihydrate difluoride
PubChem CID 24884278
SMILES O.O.[F-].[F-].[Cu++]

Specifications

Melting Point >130°C (decomposition)
Color Blue
Physical Form Crystalline
Quantity 10 g
UN Number UN3288
Merck Index 14,2638
IUPAC Name copper;difluoride;dihydrate
Formula Weight 101.54 (Anhydrous)
Percent Purity 99.99%
Grade Puratronic
Odor Odorless
Sensitivity Hygroscopic
Density 2.934g/mL
Assay (metals basis)
Chemical Name or Material Copper(II) fluoride hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.