missing translation for 'onlineSavingsMsg'
Learn More

Copper, Wire, (0.020), BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15587914 Shop All J.T.Baker Products
Click to view available options
Quantity:
454 g
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15587914

Brand: Avantor J.T.Baker 173601

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 7440-50-8
Quantity 454 g
Form Wire
Molecular Formula Cu
MDL Number MFCD00010965
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES [Cu]
IUPAC Name copper
Molecular Weight (g/mol) 63.55
PubChem CID 23978
ChEBI CHEBI:30052
Formula Weight 63.55
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Copper
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.