Learn More
Julabo™ CORIO CD-Series Heating Circulators with Open, Stainless Steel Baths and Pump Connections
BIOPHARMA
Product Code. 15430582
Click to view available options
Model:
CD-B17
CD-B39
CD-B5
Capacity (Metric):
13 to 17 L
3.5 to 5 L
35 to 41 L
Dimensions (L x W x H):
34 x 54 x 57 cm
38 x 23 x 41 cm
40 x 38 x 47 cm
Dimensions (LxWxH) Interior:
15 x 15 x 15 cm (Bath Opening)
18 x 30 x 20 cm (Bath Opening)
30 x 33 x 30 cm (Bath Opening)
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 15430582
Brand: Julabo™ 9012439
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
CORIO CD-Series Heating Circulators with Open, Stainless Steel Baths and Pump Connections are designed primarily for internal temperature control directly within the bath, however, external closed systems may be connected via the integrated pump connections.
  • Bright, White, Easy to Read Display
  • Easy to Operate Front Controls
  • Easy Switching Between Internal and External Circulation
  • Low Liquid Level Early Warning System
  • Integrated Drain Screw (except CD-B5)
  • Integrated Pump Connection (1 x M16)
  • Integrated Timer
  • PID1 and ACC Controls
  • Very Quiet
Specifications
Description Circulation Capacity Flow Rate: 15 L/min., Circulation Capacity Pressure: 0.35 bar
Capacity (Metric) 35 to 41 L
Flow Rate 15 L/min.
Temperature Range 20°C to 150°C (Working)
Type Heating Circulator with Open Bath
For Use With (Application) Internal and external applications at working temperatures up to +150°C
Heating Capacity 2 kW
Max. Pressure 0.35 bar
Model CD-B39
Temperature Stability ±0.03°C
Dimensions (L x W x H) 34 x 54 x 57 cm
Dimensions (LxWxH) Interior 30 x 33 x 30 cm (Bath Opening)