Learn More
Julabo™ CORIO CD-Series Heating Circulators with Open, Transparent Polycarbonate/Acrylic Baths and Pump Connections
BIOPHARMA
Product Code. 15430572
Click to view available options
Model:
CD-BT19
CD-BT27
Capacity (Metric):
14 to 19 L
20 to 27 L
Dimensions (L x W x H):
58 x 38 x 38 cm
58 x 38 x 43 cm
Dimensions (LxWxH) Interior:
35 x 30 x 15 cm (Bath Opening)
35 x 30 x 20 cm (Bath Opening)
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 15430572
Brand: Julabo™ 9012327
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
CORIO CD-Series Heating Circulators with Open, Transparent Polycarbonate/Acrylic Baths and Pump Connections are designed primarily for internal temperature control directly within the bath, however, external closed systems may be connected via the integrated pump connections.
  • Bright, White, Easy to Read Display
  • Easy to Operate Front Controls
  • Easy Switching Between Internal and External Circulation
  • Low Liquid Level Early Warning System
  • Class III (FL) according to DIN 12876-1
  • Integrated Pump Connection (1 x M16)
  • Integrated Timer
  • PID1 and ACC Controls
  • Very Quiet
Specifications
Description Circulation Capacity Flow Rate: 15 L/min., Circulation Capacity Pressure: 0.35 bar
Capacity (Metric) 20 to 27 L
Flow Rate 15 L/min.
Temperature Range 20°C to 100°C (Working)
Type Heating Circulator with Open Bath
For Use With (Application) Internal and external applications at working temperatures from +20°C to +100°C
Heating Capacity 2 kW
Max. Pressure 0.35 bar
Model CD-BT27
Temperature Stability ±0.03°C
Dimensions (L x W x H) 58 x 38 x 43 cm
Dimensions (LxWxH) Interior 35 x 30 x 20 cm (Bath Opening)