Learn More
Corning™ BioCoat™ Collagen I Inserts with PET Membrane
GREENER_CHOICE
Product Code. 10526001
Change view
Click to view available options
Pore Size:
0.3 μm
0.4 μm
0.45 μm
1 μm
3 μm
Type:
Cell Culture Insert
Well Plate
Quantity:
1
2
4
Unit Size:
Pack of 24
Pack of 4
Product Code. Pore Size Type Quantity unitSize
10526001 3 μm Well Plate 2 Pack of 24
10133861 0.3 μm Cell Culture Insert 1 Pack of 4
10488262 1 μm Well Plate 2 Pack of 24
10366151 0.45 μm Well Plate 4 Pack of 24
10295091 0.4 μm Well Plate 2 Pack of 24
10122851 3 μm Well Plate 4 Pack of 24
6 options
Product Code. 10526001
Brand: Corning™ 354541
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
  • Each lot is tested for the ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
  • Tested and found negative for bacteria and fungi
  • Cellware is also tested for ability to promote firm attachment of rat cerebellar granule cells
  • Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
  • Matrix components are applied as an even, optically clear coating
  • All lots tested and found negative for bacteria and fungi; cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
For research use only.
Order Info
Bulk quantities are also available; contact your Fisher Scientific Customer Service Representative.
TRUSTED_SUSTAINABILITY
Specifications
Inserts per Plate 12
Material PET
Pore Size 3 μm
Type Well Plate
For Use With (Application) Adhesion, growth and differentiation of neuronal and transfected cell lines
Quantity 2
Storage Requirements 2° to 8°C
No. of Wells 24