Learn More
Corning™ BioCoat™ Collagen I, Rat Tail Culture Dish
Click to view available options
Diameter (Metric):
35 mm
60 mm
100 mm
150 mm
Quantity:
10/cs.
100/cs.
20/cs.
40/cs.
5/cs.
Unit Size:
Pack of 10
Pack of 100
Pack of 20
Pack of 40
Pack of 5
Description
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
- All lots tested and found negative for bacteria and fungi
- Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
Specifications
| Closure Color | Clear |
| Closure Included | Closure Included |
| Closure Material | Polystyrene |
| Bottom Shape | Round |
| Color | Clear |
| Culture Area | 21.3 sq. cm |
| Material | Polystyrene |
| Shape | Round |
| Surface Treatment | Collagen Type I |
| Type | Cell Culture Dish |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur