Learn More
Corning™ BioCoat™ Control Insert, PET Membrane
Product Code. 10047911
Click to view available options
Pore Size:
0.4 μm
1 μm
3 μm
8 μm
No. of Wells:
6
24
Inserts per Plate:
6
12
Quantity:
2
4
Unit Size:
Pack of 24
Product Code. 10047911
Brand: Corning™ 354573
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Use alongside BD BioCoat™ inserts when studying effects of the ECM component present on the BD BioCoat cell culture insert
Corning BioCoat cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and/or differentiation.
Coatings include Corning Matrigel™ matrix, Fibronectin, Collagen, and Laminin.
Order Info
Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates
TRUSTED_SUSTAINABILITY
Specifications
Inserts per Plate 6
Culture Area 4.2 cm2
Material PET
Pore Size 3 μm
Type Well Plate
Quantity 4
Storage Requirements 2° to 8°C
No. per Pack 6/Pk.
No. per Case 24/Cs.
Surface Coating BioCoat Control
No. of Wells 6