Learn More
Corning™ BioCoat™ Fibronectin TC-treated Culture Dish
Click to view available options
Diameter (Metric):
35 mm
60 mm
100 mm
150 mm
Unit Size:
Pack of 10
Pack of 20
Pack of 5
Description
- Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
- Tested for their ability to promote attachment and spreading of BHK-21 hamster kidney cells
- All lots tested and found negative for bacteria and fungi
- Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
The human plasma used in preparing this product was tested and found negative for HBsAg and HIV antibody.
For adhesion, growth and differentiation of neuronal and transfected cell lines
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
Specifications
| Bottom Shape | Round |
| Color | Clear |
| Culture Area | 21.3 cm2 |
| Material | Polystyrene |
| Shape | Round |
| Type | Cell Culture Dish |
| No. per Case | 20/Cs. |
| No. per Pack | 5/Pk. |
| Diameter (Metric) | 60 mm |