Learn More
Corning™ BioCoat™ Intestinal Epithelium Differentiation Environment Kit
Shop All Corning Products
Description
Includes
- 400mL Entero-STIM™ Enterocyte Differentiation Medium (DMEM containing sodium butyrate)
- Lyophilized BD™ MITO+ Serum Extender Supplement
- 100mL Seeding Basal Medium (DMEM)
- 24 ™ Fibrillar Collagen Cell Culture Inserts (inserts contain 1μm pore size membranes packaged in one 24-well Falcon™ Companion TC Plate with one additional plate
Corning™ BioCoat™ Intestinal Epithelium Differentiation Environment is an integrated system designed to create in vitro intestinal models. The system comprises a specially formulated serum-free medium, culture supplements, sodium butyrate, and Corning BioCoat Fibrillar Collagen Cell Culture Inserts in one convenient kit.
For Customers in California only: WARNING Please refer to Safety Data Sheet (SDS) Section 15 or Letter in Lieu for more information regarding California Proposition 65.
Specifications
Specifications
| For Use With (Application) | Transport and absorption studies of drugs, metabolites and infectious diseases of the intestine |
| Storage Requirements | 2° to 8°C |
| No. per Pack | 1/Pk. |
| No. per Case | 1/Ea. |