Learn More
Corning™ BioCoat™ TC-treated Multiwell Variety Packs
Click to view available options
Item Description:
Variety pack I
Variety pack II
Unit Size:
Pack of 5
Description
- Selections include different combinations of extracellular matrix proteins and attachment factors
Multiwell Plate Variety Packs
- 6-well
- Pack I contains five plates (clear, round well, flat-bottom, TC-treated, with 9.6cm2 cell growth area)
- Coated with one of the following: Collagen I, Collagen IV, Fibronectin, Laminin, and Poly-D-Lysine
- Lids (Fisher P/N 08-774-133 only)
Specifications
Specifications
| Type | Cellware Variety Pack |
| Item Description | Variety pack I |
| Storage Requirements | 2° to 8°C |