Learn More
Corning™ Black Phenolic Threaded Screw Caps
Shop All Corning Products
Click to view available options
Neck Finish:
15-415
18-415
28-410
38-430
40-400
No. per Pack:
12/Pk.
288/Pk.
32/Pk.
72/Pk.
Unit Size:
Pack of 12
Pack of 192
Pack of 288
Pack of 72
Description
These caps are fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving. Caps have a glued-in, white rubber liner.
Specifications
Specifications
| Color | Black |
| Material | Phenolic Resin |
| No. per Pack | 12/Pk. |
| Liner Material | Rubber |
| Neck Finish | 38-430 |
| Height (Metric) | 27 mm |
| Product Type | Screw Cap |