missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Ciprofloxacin Hydrochloride

Packaged in plastic bottles

Brand:  Corning™ 61-277-RF

Additional Details : CAS : 77-86-1 Poids : 0.01000kg

 View more versions of this product

Product Code. 15383771

  • 149.00€ / 1g


En stock, prêt à être expédié : 1
Add to basket

Specifications

Specifications

Powder
Plastic Bottle
15° to 30°C
1g
Ciprofloxacin Hydrochloride
15° to 30°C
Videos
Documents