Learn More
Corning™ X-GAL and IPTG, Powder
Produced under highest industry standards
Brand:  Corning™ 46-101-RF
Additional Details : Poids : 0.01000kg
 View more versions of this product
Product Code. 15383721
  • 272.00€ / 1g
Expédition estimée
12-11-2020
Add to basket
Description
Description
X-Gal (5-bromo-4-chloro-3-indolyl-b-D-galactopyranoside)
  • Substrate of b-galactosidase
  • Hydrolyzed by β-galactosidase to produce indoxyl in bacterial medium
IPTG (isopropylthiogalactoside)
  • Inducer of lac operon in bacteria
  • Used with X-gal for detecting lac gene activity
  • For screening of clones containing gene of interest in lac operon
Specifications
Specifications
1000 mg
5-bromo-4-chloro-3-indolyl-b-D-galactopyranoside
Research use only