Learn More
Corning™ DeckWorks™ Low-Binding Hinged Rack Pipet Tips, Nonsterile
Shop All Corning ProductsDescription
Corning DeckWorks low binding tips are available in 10 μL, 200 μL, and 300 μL sizes. Proprietary low binding technology reduces surface tension on the interior wall of the tip. Leaching and/or subsequent sample degradation associated with silicone-based tips are eliminated. DeckWorks low binding tips are ideal for quantitative assays in which sample binding can significantly alter results. Specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins and/or peptides.
Specifications
Specifications
| Volume (Metric) | 1 to 300 μL |
| Sterility | Non-sterile |
| Color | Natural |
| For Use With (Application) | Designed for quantitative assays in which sample binding can significantly alter results |
| Format | Hinged Rack |
| Tip Style | Low Retention, Round |
| Packaging | Hinged rack |
| For Use With (Equipment) | Universal Fit |
| Graduated | Yes |
| Product Line | DeckWorks |
| Show More |