Learn More
Corning™ Flexible Packaging Systems: MPC Connector Tubing
Clear tubing; female MPC connector
Brand: Corning™ 91-700-12
Additional Details : Weight : 0.42750kg
Description
- Chemically resistant
- Heat sealable
- Flexible
Specifications
Connector tubing | |
cGMPs, FDA Regulation 21 CFR part 820 and ISO 13485 Quality | |
Pinch clamp and filling bell, 0.25 in. barb |
Fits tubing 60.96 cm | |
Clear |