Learn More
Corning™ Intestinal Epithelium Differentiation Medium
GREENER_CHOICE
Product Code. 11553600 Shop All Corning Products
Click to view available options
Quantity:
1 x 250 mL
Unit Size:
500mL
Product Code. 11553600
Brand: Corning™ 355357
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Promotes the rapid differentiation of intestinal epithelium in vitro
Corning™ Intestinal Epithelium Differentiation Medium (formerly Entero-STIM), 250 mL, promotes the rapid differentiation of intestinal epithelium in vitro, 2/pack (same as Cat. #30357M)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stem Cell Differentiation Media
Sterility Sterile
Quantity 1 x 250 mL
Content And Storage 2° to 8°C