Learn More
Corning™ LSE™ Vortex Mixer
GREENER_CHOICE
Product Code. 11931991 Shop All Corning Products
Click to view available options
Voltage:
120 V
Unit Size:
Each
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Product Code. 11931991
Brand: Corning™ 6775
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Includes
Cup head for all standard microtubes and centrifuge tubes
Variable speed mixer provides fast, efficient mixing with minimal vibration
The Corning LSE variable speed vortex mixer provides fast, efficient mixing with minimal vibration. The unit features a powerful, counter-balanced motor drive, and continuous or 'touch' mix modes. A range of accessories are available for all common sample tubes and containers.
  • Variable speed for gentle to vigorous mixing
  • Optimized counter balance for minimal vibration
  • Lightweight and portable
  • Includes cup head for all standard microtubes and centrifuge tubes
  • Mix a variety of tubes and plates with optional accessory heads
  • Touch and continuous mix modes
TRUSTED_SUSTAINABILITY
Specifications
Speed Range 0 to 3400 rpm
Electrical Requirements 120 V, 50/60 Hz
Voltage 120 V
Product Type Vortex Mixer
Frequency 50/60 Hz
Height (English) 5.11 in.
Height (Metric) 13 cm
Length (English) 5.51 in.
Length (Metric) 14 cm
Width (English) 6.29 in.
Width (Metric) 16 cm
Weight (English) 4.8 lb.
Weight (Metric) 2.2 kg