Learn More
Corning™ PureCoat™ Carboxyl Surface Dishes
Negatively charged surface improves attachment. increases proliferation and improved differentiation for a broad range of cells that exhibit poor attachment properties in serum-free or serum-reduced conditions
48.71€ - 154.19€
Specifications
Bottom Shape | Flat |
---|---|
Culture Area | 58.1 cm2 |
Material | Polystyrene |
Shape | Round |
Surface Treatment | Carboxyl |
Product Code | Brand | No. per Case | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. per Case | Price | Quantity & Availability | |||||
11503590
|
Corning™
354784 |
10/Cs. |
48.71€
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
11553640
|
Corning™
356784 |
40/Cs. |
154.19€
Pack of 40 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
Description
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Specifications
Flat | |
Polystyrene | |
Carboxyl | |
10/Pk. |
58.1 cm2 | |
Round | |
Cell Culture Dish | |
100 mm |