missing translation for 'onlineSavingsMsg'
Learn More

Cortisone 21-acetate, 98+%, Acros Organics

Brand:  Acros Organics 448960250

Additional Details : CAS : 50-04-4 Weight : 0.03000kg


Packaging Glass bottle
Quantity 25g

 View more versions of this product

Product Code. 13581610

  • 353.00€ / 25g


In Stock 1
Add to basket


Specifications

Chemical Identifiers

50-04-4
402.48
cortisone acetate, cortisone 21-acetate, cortone acetate, incortin, biocort acetate, artriona, cortadren, scheroson, cortisyl, adreson
CHEBI:3897
CC(=O)OCC(=O)C1(CCC2C1(CC(=O)C3C2CCC4=CC(=O)CCC34C)C)O
C23H30O6
ITRJWOMZKQRYTA-RFZYENFJSA-N
5745
[2-[(8S,9S,10R,13S,14S,17R)-17-hydroxy-10,13-dimethyl-3,11-dioxo-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate

Specifications

Cortisone 21-acetate
237°C to 240°C
50-04-4
C23H30O6
cortisone acetate, cortisone 21-acetate, cortone acetate, incortin, biocort acetate, artriona, cortadren, scheroson, cortisyl, adreson
CC(=O)OCC(=O)C1(CCC2C1(CC(=O)C3C2CCC4=CC(=O)CCC34C)C)O
402.48
CHEBI:3897
98+%
Glass bottle
25g
98% min. (HPLC)
15,2525
ITRJWOMZKQRYTA-RFZYENFJSA-N
[2-[(8S,9S,10R,13S,14S,17R)-17-hydroxy-10,13-dimethyl-3,11-dioxo-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
5745
402.48
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Cortisone 21-acetate

Signal Word
  • Warning

Hazard Category
  • Reproductive toxicity Category 2

Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Documents