missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Cotinine.HCl, Lipomed
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
(-)-Cotinine.HCl
Specifications
Specifications
| Chemical Name or Material | (-)-Cotinine Hydrochloride |
| CAS | 486-56-6 (FB) |
| Analyte or Component Names | (-)-Cotinine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H12 N2 O . Cl H |
| Synonym | (-)-Cotinine HCl, (5 S)-1-Methyl-5-(3-pyridyl)-pyrrolidin-2-one hydrochloride |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.CN1[C@@H](CCC1=O)c2cccnc2 |
| IUPAC Name | (5 S)-1-methyl-5-pyridin-3-ylpyrrolidin-2-one;hydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction