Learn More
DuPont™ Tyvek 200 Easysafe Hooded Coverall
5.20 EUR valid until 2024-12-31
Use promo code "21990" to get your promotional price.
3.25 EUR valid until 2024-04-30
Use promo code "21637" to get your promotional price.
Use promo code "21990" to get your promotional price.
3.25 EUR valid until 2024-04-30
Use promo code "21637" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Robust yet lightweight
- Elasticated face, wrists, waist and ankles
- Antistatic treatment on the outside
- Stitched external seams in yellow for visual identification and differentiation
Specifications
Product Type | Hooded Coveralls |
Size | 3X-Large |
Material | Polyethylene |
Color | White |
Ankle Style | Elastic |
Neck Style | Hooded |
Hood Type | 2-panel |
Closure Type | Zipper |
Seam Type | Stitched |
Protection Class | Category III, Type 5 and 6 |