Learn More
Tocris Bioscience™ Covidcil-19
GREENER_CHOICE
Product Code. 18739256
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18739256
Brand: Tocris Bioscience™ 7485/10
Binds to SARS-CoV-2 frameshifting element and reduces frameshifting efficiency
Covidcil-19 binds to the revised attenuator hairpin structure of the SARS-CoV-2 frameshifting element (FSE) with high affinity (Kd = 11 nM). It stabilizes the hairpin's folded state and reduces frameshifting efficiency in cells. Covidcil-19 inhibits viral propagation and reduces viral infectivity by > 3.5 orders of magnitude.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-[[4-(Methylamino)-2-quinazolinyl]amino]benzoic acid
CAS 1225177-95-6
Quantity 10 mg
Target Antiviral Compounds
Molecular Formula C16H14N4O2
Purity 0.98