missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ COX2 Rabbit Monoclonal Antibody

Obtain accurate, reproducible results in Immunohistochemistry experiments with Epredia™ COX2, Rabbit Monoclonal Antibody, part of a comprehensive antibody portfolio.

167.28€ - 700.00€

Specifications

Antigen COX2
Clone SP21
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Form Quantity Price Quantity & Availability  
Product Code Brand Form Quantity Price Quantity & Availability  
12603777
View Documents Promotion Details
Epredia™
RM-9121-S0
Concentrated 100 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12613777
View Documents Promotion Details
Epredia™
RM-9121-S1
Concentrated 500 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12693767
View Documents Promotion Details
Epredia™
RM-9121-S
Concentrated 1 mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12673767
View Documents Promotion Details
Epredia™
RM-9121-R7
Prediluted 7 mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12683767
View Documents Promotion Details
Epredia™
RM-9121-RQ
Ready-to-Use 12 mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

COX2 (Cyclooxygenase-2) is an inducible enzyme. It is involved in the response of cells to growth factors, tumor promoters, and cytokines that induce its expression. Given its role in synthesizing prostaglandins, COX2 is therefore of interest in studying immune response regulation. COX2 is induced by a wide variety of stimuli and was initially identified as immediate-early growth response gene. In addition, COX2 expression markedly increased in 85-90% of human colorectal adenocarcinoma whereas COX1 levels remain unchanged.

Host Species: Rabbit

Clone: SP21

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide from C-terminus of rat COX2

Molecular Weight: 70 kDa

Positive Control: Lung and colon CAs

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

COX2
Immunohistochemistry (Paraffin)
Unconjugated
IVD
A synthetic peptide from C-terminus of rat cox2
SP21
Monoclonal
Rabbit
Enzyme
Primary
Videos
SDS
Documents

Documents

Product Certifications